Order Entry
Austria
Orders LinkContactUsLinkComponent
Methylcyclohexane ≥99%

Methylcyclohexane ≥99%

Glass bottle for solids, 100 g
Catalog # 31861.22
Flammable
Health
Irritant
Environment
Methylcyclohexane ≥99%
Catalog # 31861.22
CAS Number:  108-87-2
Methylcyclohexane ≥99%
Methylcyclohexane ≥99%

Glass bottle for solids, 100 g

CAS Number:  108-87-2
Price
69,60 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99+% CAS No.: 108-87-2 Molecular Formula: C7H14 Formula Weight: 98.19 Flash Point: -4°(-25°F) Physical Form: Liquid Density: 0.771 Refractive Index: 1.4230 Appearance: Colourless Melting Point: -126° Boiling Point: 101° Beilstein Reference: 505972 Merck Reference: 14,6047 UN No.: 2296 MDL No.: MFCD00001497

  • CAS Number:108-87-2
  • Formula:C₆H₁₁CH₃
  • MDL Number:MFCD00001497
  • Molecular Weight:98.19 g/mol
  • Merck Index:14,6047
  • Boiling Pt:101 °C
  • Melting Pt:–126 °C
  • Density:0.771
  • Flash Pt:–4 °C (–25 °F)
  • Storage Temperature:Ambient
  • EINECS:203-624-3
  • UN:2296
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications