Orders LinkContactUsLinkComponent
Microscope adapters for micromanipulators
Microscope adapters for micromanipulators
Nikon Eclipse® TS100
Catalog # 631-0217
Supplier:   EPPENDORF
Microscope adapters for micromanipulators
Catalog # 631-0217
Supplier:  EPPENDORF
Microscope adapters for micromanipulators
Microscope adapters for micromanipulators
Nikon Eclipse® TS100
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Microscope adapters for InjectMan® NI 2 and TransferMan® NK 2. The universal stand is suitable for mounting manipulators on upright or stereo microscopes, independent of the microscope tripod used.
Specifications
  • Description:
    Nikon Eclipse® TS100