Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Objektträgerkästen, SlideFile™

Pink, PS, 100, 82×245×82 mm
  631-1902
Objektträgerkästen, SlideFile™
  631-1902
Objektträgerkästen, SlideFile™
Objektträgerkästen, SlideFile™

Pink, PS, 100, 82×245×82 mm

 
235,00 €
   
1 * 10 ST
 

This impact resistant storage system can hold up to 400 slides per unit, for space efficient storage. All slides are stored upright for easier manipulation.

  • Removable tray inside the storage box for 100 individual numbered slots
  • Transparent hinged cover makes it easy to see the contents of the box at a glance
  • Unique feature with this system to read barcodes without having to remove the slides from the box

W×D×H: 82×245×86 mm

 : Each system includes a slide box and a removable tray.

 : Index cards numbered from one to 100 for recording slide titles are supplied with each box.

 
  • Anzahl Objektträger:
    100
  • Farbe:
    Pink
  • Material:
    PS
  • B×T×H:
    82×245×82 mm