Accessories for analytical Mill, A 11 basic
Beater A11.3, Pulverising substances with a Mohs hardness of up to 9
Catalog # 412-1102
Supplier: IKA
Jump to:
Product Details & Documents
Specifications
- Description:Beater A11.3
- Seq:04
- For:Pulverising substances with a Mohs hardness of up to 9