REFRIGERATOR FORCED AIR COOLING 174L HEIGHT 884 X WIDTH 602 X DEPTH 600 MM 3 SHELVES AND 1 GRID 1 * 1 items
472-2203
: Liebherr
REFRIGERATOR FORCED AIR COOLING 174L HEIGHT 884 X WIDTH 602 X DEPTH 600 MM 3 SHELVES AND 1 GRID 1 * 1 items
472-2203
: Liebherr
Frequently Bought Together
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection