Orders LinkContactUsLinkComponent
Container, Securitainer®
Container, Securitainer®
Securitainer®-cap, 62 mm
Catalog # COVEC62/01
Supplier:   COVERA
Container, Securitainer®
Catalog # COVEC62/01
Supplier:  COVERA
Container, Securitainer®
Container, Securitainer®
Securitainer®-cap, 62 mm
Price
626,00 €
Log in for availability
1 * 900 ST
Product Details & Documents
SECURITAINER®, made of white PP is an effective security container. It has two main uses: as an individual transport container for glass vials and as packaging for materials and substances sensitive to dampness.
  • The tamper-proof seal guarantees that the packaging has not been opened-providing increased product security
  • The seal prevents manipulation of vials, labels and their contents
  • The strong, light construction protects the contents effectively
  • Easy to handle
  • Lids to be ordered seperately
Specifications
  • Cap Size:
    62 mm
  • Description:
    Securitainer®-cap
  • Seq:
    14