Orders LinkContactUsLinkComponent
Microtubes, Prepared
Microtubes, Prepared
Microtubes, prepared, glucose fluoride, with attached push cap closure, 1,3 ml, 1000/case
Artikelnr. SARS41.1505.005
Marketsource
Anbieter:   SARSTEDT
Microtubes, Prepared
Artikelnr. SARS41.1505.005
Anbieter:  SARSTEDT
Marketsource
Microtubes, Prepared
Microtubes, prepared, glucose fluoride, with attached push cap closure, 1,3 ml, 1000/case
Preis
148,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
PP, translucent with PP screw cap.
  • Type of preparation: Trisodium citrate solution
  • With paper label
  • Graduated
  • Recommended centrifugation: 1800×g
With screw cap or push-on cap.
Specifications
  • Packed:
    1000/case
  • Capacity:
    1,3 ml
  • Description:
    Microtubes, prepared, glucose fluoride, with attached push cap closure
  • Seq:
    8