Order Entry
Austria
Orders LinkContactUsLinkComponent
Spoon spatulas, micro

Spoon spatulas, micro

130 mm, 5 mm
Catalog # 231-2151
Supplier:   RSG
Spoon spatulas, micro
Catalog # 231-2151
Supplier:  RSG
Spoon spatulas, micro
Spoon spatulas, micro

130 mm, 5 mm

Price
4,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel 18/10, anti-magnetic.

  • With round handle and one bent spoon
  • Excellent temperature resistance, autoclavable
  • Good resistance to chemicals
  • Very flexible

Documents

Specifications
  • Length:
    130 mm
  • Blade width:
    5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.