Order Entry
Austria
Orders LinkContactUsLinkComponent
Scoops

Scoops

1000 ml, 320 mm
Catalog # 231-0203
Supplier:   RSG
Scoops
Catalog # 231-0203
Supplier:  RSG
Scoops
Scoops

1000 ml, 320 mm

Price
34,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel. Manual scoops for collecting or measuring powders, pastes or granulates.

  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Robust design

Documents

Specifications
  • Capacity:
    1000 ml
  • Length:
    320 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.