Scoops
120 ml, 190 mm
Catalog # 231-0201
Supplier: RSG
Jump to:
Product Details & Documents
Stainless steel. Manual scoops for collecting or measuring powders, pastes or granulates.
- Excellent thermal resistance, autoclavable
- Good chemical resistance
- Robust design
Dokumente
Certificates
Search by Lot NumberSpecifications
- Capacity:120 ml
- Length:190 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



