Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Registrieren
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Scissors
Microscopy scissors
Microscopy scissors
Print
Microscopy scissors
Curved, sharp/sharp, 100 mm
Catalog # 233-1454
Supplier:
RSG
Microscopy scissors
Catalog # 233-1454
Supplier:
RSG
Expand image
Return to Top
Microscopy scissors
Curved, sharp/sharp, 100 mm
Specifications
Add to cart
Jump to:
Specifications
Price
20,30 €
Log in
for availability
Unit of Measure
1 * 1 ST
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Stainless steel.
Warm forged
Corrosion resistant
Specifications
Type:
Curved, sharp/sharp
Length:
100 mm
Formalin 10%, Q Path®, buffered
#: FOR0150AF59001
Supplier: VWR Chemicals