Orders LinkContactUsLinkComponent
Microscopy scissors
Microscopy scissors
Straight, sharp/sharp, 90 mm
Catalog # 233-1551
Supplier:   RSG
Microscopy scissors
Catalog # 233-1551
Supplier:  RSG
Microscopy scissors
Microscopy scissors
Straight, sharp/sharp, 90 mm
Price
22,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel.
  • Warm forged
  • Corrosion resistant
Specifications
  • Type:
    Straight, sharp/sharp
  • Length:
    90 mm