Order Entry
Austria
Orders LinkContactUsLinkComponent
Sample loops for sample injectors, Rheodyne®

Sample loops for sample injectors, Rheodyne®

Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100, Stainless steel, 50 µl, 0,51 mm, 1,5 mm
Catalog # 554-2158
Supplier:   IDEX RHEODYNE
Sample loops for sample injectors, Rheodyne®
Catalog # 554-2158
Supplier:  IDEX RHEODYNE
Sample loops for sample injectors, Rheodyne®
Sample loops for sample injectors, Rheodyne®

Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100, Stainless steel, 50 µl, 0,51 mm, 1,5 mm

Price
120,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Capacity:
    50 µl
  • Description:
    Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100
  • Material:
    Stainless steel
  • Seq:
    25
  • For tube I-Ø:
    0,51 mm
  • For tube O-Ø:
    1,5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.