Order Entry
Austria
Orders LinkContactUsLinkComponent
Formaldehyde 35% in aqueous solution stabilised, TECHNICAL

Formaldehyde 35% in aqueous solution stabilised, TECHNICAL

Plastic drum, 10 L
Catalog # 90240.9010
Health
Toxic
Corrosive
Supplier:   VWR Chemicals
Formaldehyde 35% in aqueous solution stabilised, TECHNICAL
Catalog # 90240.9010
Supplier:  VWR Chemicals
CAS Number:  50-00-0
Formaldehyde 35% in aqueous solution stabilised, TECHNICAL
Formaldehyde 35% in aqueous solution stabilised, TECHNICAL

Plastic drum, 10 L

CAS Number:  50-00-0
Price
160,00 €
Log in for availability
1 * 10 L
Product Details & Documents

Warning : Store this product at ambient temperature (above 15°C), because it may polymerise at temperatures below 9°C. Presence of polymers does not affect product properties. Precipitate can be removed by filtration.

Synonyms:  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Density:1.088 g/cm³ (20 °C)
  • Flash Pt:66 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-001-8
  • UN:2209
  • ADR:8,III
  •  :Stabilised with methanol 10-15 %
Specifications

Specification Test Results

  • Assay:
    Min. 34.5 %

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    10 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.