Blind small flanges
Blanking flange, DN 10, Stainless steel
185-2111
Blind small flanges KF made of aluminium or stainless steel.
Used for vacuum system.
- Description:Blanking flange, DN 10
- Material:Stainless steel
- Seq:06