Orders LinkContactUsLinkComponent
Blind small flanges
Blind small flanges
Blanking flange, DN 40, Aluminium
Catalog # 185-2104
Supplier:   VACUUBRAND
Blind small flanges
Catalog # 185-2104
Supplier:  VACUUBRAND
Blind small flanges
Blanking flange, DN 40, Aluminium
Price
18,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
Blind small flanges KF made of aluminium or stainless steel.
Used for vacuum system.
Documents
Specifications
  • Description:
    Blanking flange, DN 40
  • Material:
    Aluminium
  • Seq:
    04