Orders LinkContactUsLinkComponent
Screwthread joints for VACUU.LAN®
Elbow (90°) adapter, for G 3/8" thread to PTFE tubing 10/8 mm
Catalog # VACU20639854
Supplier:   VACUUBRAND
Screwthread joints for VACUU.LAN®
Catalog # VACU20639854
Supplier:  VACUUBRAND
Screwthread joints for VACUU.LAN®
Elbow (90°) adapter, for G 3/8" thread to PTFE tubing 10/8 mm
Price
21,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
Adapters for VACUU.LAN® local area vacuum network components.
Specifications
  • Description:
    Elbow (90°) adapter, for G 3/8" thread to PTFE tubing 10/8 mm
  • Seq:
    02