Order Entry
Austria
Orders LinkContactUsLinkComponent
illustra™ NICK™ columns

illustra™ NICK™ columns

Illustra™ NICK™ columns, 50
Catalog # 17-0855-02
Supplier:   Cytiva
illustra™ NICK™ columns
Catalog # 17-0855-02
Supplier:  Cytiva
CAS Number:  
illustra™ NICK™ columns
illustra™ NICK™ columns

Illustra™ NICK™ columns, 50

CAS Number:  
Price
542,00 €
Log in for availability
1 * 50 ST
Product Details & Documents

NICK™ columns were originally designed for purification of nick-translated DNA fragments but are also suitable for separation of any labelled probe from unincorporated labelled nucleotides.

  • For rapid purification of labeled DNA (20 bases in length) from unincorporated radiolabeled nucleotides using gravity-flow chromatography
  • Prepacked with Sephadex™ G-50 DNA Grade in distilled water with 0,15% Kathon CG/ICP Biocide
  • Can accommodate sample volumes up to 100 μl

They are ready for immediate use, and operate by gravity flow. Tests show that at least 97% of radioactivity applied in a nick-translated mixture elutes in two well-separated peaks, corresponding to labeled DNA and unincorporated nucleotides, respectively. At least 90% of the applied DNA is recovered.

Specifications

Specifications

  • Pk:
    50
  • Description:
    Illustra™ NICK™ columns
  • Cat. No.:
    17-0855-02
  • Prep size:
    Scalable
  • Refine target molecule:
    Amplified/modified DNA
  • Target molecule:
    DNA
  • Method / Format:
    Gel filtration
  • Description:
    Illustra™ NICK™ columns
  • Sample type:
    PCR and other enzymatic reactions

Specifications