Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Dissecting needles

Lancet, 140 mm
  USBE5490
Dissecting needles
  USBE5490
Dissecting needles
Dissecting needles

Lancet, 140 mm

 
2,40 €
   
1 * 1 ST
 

Stainless steel needles with plastic handles.

Weight: 4 - 5 g

 
  • Form:
    Lancet
  • Length:
    140 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.