Order Entry
Austria
Orders LinkContactUsLinkComponent
Scoops

Scoops

2500 ml, 410 mm
Catalog # USBE3727
Supplier:   USBECK Laborgeräte
Scoops
Catalog # USBE3727
Scoops
Scoops

2500 ml, 410 mm

Price
37,80 €
Log in for availability
1 * 1 ST
Product Details & Documents

Aluminium, smooth. For collecting or measuring powders, granulates or pastes.

  • Good chemical resistance
  • Excellent thermal resistance
  • Light, robust and easy to use
Specifications
  • Capacity:
    2500 ml
  • Length:
    410 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.