Order Entry
Austria
Orders LinkContactUsLinkComponent
Scoops

Scoops

250 ml, 250 mm
Catalog # 231-0325
Supplier:   USBECK Laborgeräte
Scoops
Catalog # 231-0325
Scoops
Scoops

250 ml, 250 mm

Price
7,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Aluminium, smooth. For collecting or measuring powders, granulates or pastes.

  • Good chemical resistance
  • Excellent thermal resistance
  • Light, robust and easy to use
Specifications
  • Capacity:
    250 ml
  • Length:
    250 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.