Orders LinkContactUsLinkComponent
Blunt forceps
Blunt forceps
Tweezers, 145 mm
Catalog # 232-0105
Supplier:   USBECK Laborgeräte
Blunt forceps
Catalog # 232-0105
Blunt forceps
Blunt forceps
Tweezers, 145 mm
Price
5,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel.
  • Curved
  • Blunt pointed ends
Specifications
  • Description:
    Tweezers
  • Length:
    145 mm