Order Entry
Austria
Orders LinkContactUsLinkComponent
Stand rods

Stand rods

12 mm, 750 mm
Catalog # 241-7152
Supplier:   USBECK Laborgeräte
Stand rods
Catalog # 241-7152
Stand rods
Stand rods

12 mm, 750 mm

Price
10,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Steel, galvanised.

  • Good chemical resistance
  • Very sturdy
Specifications
  • Ø:
    12 mm
  • Length:
    750 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.