Stand bases
250×160 mm, 2,5 kg
241-7175
Stainless steel. Firm, non slip stand for a wide variety of uses. With an M10 threaded hole on face side and adjustable feet for uneven surfaces.
- Rubber feet for a firm footing
- Smooth surfaces for easy, fast cleaning
- Robust
DIN 12892/55
: DIN 12892/55
- L×W:250×160 mm
- Seq:03
- Weight:2,5 kg



