Order Entry
Austria
Orders LinkContactUsLinkComponent
Stand plates

Stand plates

315×200 mm, 4,0 kg
Catalog # 241-7113
Supplier:   USBECK Laborgeräte
Stand plates
Catalog # 241-7113
Stand plates
Stand plates

315×200 mm, 4,0 kg

Price
41,30 €
Log in for availability
1 * 1 ST
Product Details & Documents

Steel, powder coated. Firm, non slip stand. For a wide variety of uses. With M10 threaded hole on face side.

  • Rubber feet for a firm footing
  • Easy to clean
  • Robust
Specifications
  • L×W:
    315×200 mm
  • Weight:
    4,0 kg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.