Order Entry
Austria
Orders LinkContactUsLinkComponent
Stand plates

Stand plates

180×100 mm, 1,1 kg
Catalog # 241-7110
Supplier:   USBECK Laborgeräte
Stand plates
Catalog # 241-7110
Stand plates
Stand plates

180×100 mm, 1,1 kg

Price
17,30 €
Log in for availability
1 * 1 ST
Product Details & Documents

Steel, powder coated. Firm, non slip stand. For a wide variety of uses. With M10 threaded hole on face side.

  • Rubber feet for a firm footing
  • Easy to clean
  • Robust
Specifications
  • L×W:
    180×100 mm
  • Weight:
    1,1 kg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.