Orders LinkContactUsLinkComponent
Inlet liners
Inlet liners
Silanized Glass Wool
Catalog # PERC03300905
Supplier:   PerkinElmer
Inlet liners
Catalog # PERC03300905
Supplier:  PerkinElmer
Inlet liners
Silanized Glass Wool
Price
104,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Liners protect GC columns against column contamination. These glass split and splitless inlet liners promote full vaporisation of samples, minimise mass discrimination, and achieve accurate and reproducible results.
  • Liners packed with wool help to trap non-volatile material
Specifications
  • Description:
    Silanized Glass Wool
  • Seq:
    09