Orders LinkContactUsLinkComponent
Affinity protein expression and purification systems
Calmodulin affinity resin, Calmodulin binding peptide (CBP) affinity resin, 10 mL
Catalog # STRA214303
Marketsource
Supplier:   STRATAGENE
Affinity protein expression and purification systems
Catalog # STRA214303
Supplier:  STRATAGENE
Marketsource
CAS Number:  
Affinity protein expression and purification systems
Calmodulin affinity resin, Calmodulin binding peptide (CBP) affinity resin, 10 mL
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 10 mL
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
The affinity protein expression and purification system provides a simple, gentle and effective method for high-level protein expression and purification.
  • pCAL vectors derived from T7 RNA polymerase-based pET vectors
  • Novel purification tag combines small size, gentle elution and high-affinity binding
  • Easy tag removal resulting in native protein
 : The complete system includes pCAL expression vectors containing a calmodulin-binding peptide tag and calmodulin affinity resin (available separately).
 : For research use only. Not for use in diagnostic procedures.
Specifications
Specifications
  • Pk:
    10 mL
  • Description:
    Calmodulin affinity resin
  • Details:
    Calmodulin binding peptide (CBP) affinity resin
Specifications