Orders LinkContactUsLinkComponent
Containers with optional snap caps, Nunc™
Containers with optional snap caps, Nunc™
for 50 ml containers, Grey
Catalog # 391-8581
   Low Manufacturing Impact
Containers with optional snap caps, Nunc™
Catalog # 391-8581
   Low Manufacturing Impact
Containers with optional snap caps, Nunc™
Containers with optional snap caps, Nunc™
for 50 ml containers, Grey
Price
520,00 €
Log in for availability
1 * 2.400 ST
Product Details & Documents
PS, clear.
  • Transparent polystyrene construction
  • Versatile containers made of high-quality resins
  • Snap caps available for all containers
  • Cylindrical shape
  • Non sterile
Ordering information: Caps must be ordered seperately.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
  • Type:
    for 50 ml containers
  • Cap colour:
    Grey
  • Cat. No.:
    391-8581
  • Environmental certification:
    ISO 14001 (Certificate Registration No.CERT-IT000330)
  • Environmentally Preferable:
    Low Manufacturing Impact