Orders LinkContactUsLinkComponent
Vectors, pBlueScript II
VCSM13 interference resistant helper phage, Interference-resistant helper phage (6 kB, single stranded). Derived from M13 K07 mutant. Carries kanamycin-resistance gene in IG region. High rescue efficiencies with kanamycin selection, size: 1 ml, 1
Catalog # STRA200251
Marketsource
Supplier:   STRATAGENE
Vectors, pBlueScript II
Catalog # STRA200251
Supplier:  STRATAGENE
Marketsource
CAS Number:  
Vectors, pBlueScript II
VCSM13 interference resistant helper phage, Interference-resistant helper phage (6 kB, single stranded). Derived from M13 K07 mutant. Carries kanamycin-resistance gene in IG region. High rescue efficiencies with kanamycin selection, size: 1 ml, 1
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 ST
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
The pBluescript II phagemids (plasmids with a phage origin) are cloning vectors designed to simplify commonly used cloning and sequencing procedures, including the construction of nested deletions for DNA sequencing, generation of RNA transcripts in vitro and site-specific mutagenesis and gene mapping.
  • High copy number ColE1-based phagemid
  • Large and versatile polylinker in two orientations, f1 origin available in either (+) or (–) orientation
  • T3 and T7 promoters for in vitro transcription of RNA
The pBluescript II phagemids have an extensive polylinker with 21 unique restriction enzyme recognition sites.
 : For research use only. Not for use in diagnostic procedures.
Specifications
Specifications
  • Pk:
    1
  • Description:
    VCSM13 interference resistant helper phage, Interference-resistant helper phage (6 kB, single stranded). Derived from M13 K07 mutant. Carries kanamycin-resistance gene in IG region. High rescue efficiencies with kanamycin selection, size: 1 ml
Specifications