Orders LinkContactUsLinkComponent
Microscope slides, Nunc™
Microscope slides, Nunc™
Microscope slides, sterile, PS, 25×75 mm
Catalog # 734-2070
   Low Manufacturing Impact
Microscope slides, Nunc™
Catalog # 734-2070
   Low Manufacturing Impact
Microscope slides, Nunc™
Microscope slides, Nunc™
Microscope slides, sterile, PS, 25×75 mm
Price
664,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
These microscope slides are designed for applications in cell culture.
The surface treatment ensures optimal conditions for cell attachment and growth.
Specifications
  • Description:
    Microscope slides, sterile, PS
  • L×W:
    25×75 mm
  • Cat. No.:
    734-2070
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact