Order Entry
Austria
Orders LinkContactUsLinkComponent
Extraction thimbles

Extraction thimbles

33×118 mm, 1,5 mm
Catalog # 533-0010
Extraction thimbles
Catalog # 533-0010
Extraction thimbles
Extraction thimbles

33×118 mm, 1,5 mm

Price
155,00 €
Log in for availability
1 * 25 ST
Product Details & Documents

Grade 603 standard cellulose extraction thimbles, made of high quality cellulose, are available in a range of sizes suitable for use in thimble holders and automated extraction apparatus to ensure optimal fit.

Dokumente

Specifications
  • Int.Ø×H:
    33×118 mm
  • Seq:
    19
  • Thickness:
    1,5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.