ContactUsLinkComponent
FRACTOGEL EMD DEAE (S) WEAK ANION EXCHANGER, SUSPENSION IN 20% ETHANOL AND 150 MM NACL (20-40 MYM) This product cannot be returned a fter delivery 1 * 10 ml
Catalog # 1.16888.0010
FRACTOGEL EMD DEAE (S) WEAK ANION EXCHANGER, SUSPENSION IN 20% ETHANOL AND 150 MM NACL (20-40 MYM) This product cannot be returned a fter delivery 1 * 10 ml
Catalog # 1.16888.0010
Frequently Bought Together