Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Bath fluids, Supelco®

Heating bath fluid, ≤170 °C, ~330 cP, 2,5 l
Catalog # 1.15265.2500
Supplier:   Merck
Bath fluids, Supelco®
Catalog # 1.15265.2500
Supplier:  Merck
CAS Number:  
Bath fluids, Supelco®

Heating bath fluid, ≤170 °C, ~330 cP, 2,5 l

CAS Number:  
Price
321,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents

Bath fluids for improved thermostat life.

  • Oil bath fluid can be used up to approximately 250 °C
  • Heating bath fluid can be used up to approximately 170 °C
  • Non corrosive
  • High flash point
  • Low evaporation

Note: At temperatures approaching the recommended maximum operating temperature it may be advisable to use these products in a fume cupboard or vented area.

 : Note: At temperatures approaching the recommended maximum operating temperature it may be advisable to use these products in a fume cupboard or vented area.
1 cP = 1 mPa.s

Specifications

Specifications

  • Pk:
    2,5 l
  • Description:
    Heating bath fluid
  • Temperature range:
    ≤170 °C
  • Viscosity:
    ~330 cP
  • Cat. No.:
    1.15265.2500
  • M-Clarity quality level:
    MQ100

Specifications