Orders LinkContactUsLinkComponent
Test kits, COD cell test, Spectroquant®, Supelco®
Test kits, COD cell test, Spectroquant®, Supelco®
COD cell test*7-1, 10 - 150 mg/L, Chromosulphuric oxidation, Prove, Nova 30/60, Multy, Move, Picco, 25 Tests
Catalog # 1.14540.0001
Supplier:   Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Catalog # 1.14540.0001
Supplier:  Merck
CAS Number:  
Test kits, COD cell test, Spectroquant®, Supelco®
Test kits, COD cell test, Spectroquant®, Supelco®
COD cell test*7-1, 10 - 150 mg/L, Chromosulphuric oxidation, Prove, Nova 30/60, Multy, Move, Picco, 25 Tests
CAS Number:  
Price
174,00 €
Log in for availability
1 * 25 Tests
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*7 This method is officially recognised by the USEPA as an alternative method for the investigation of the following:
*7-1 wastewater
*7-2 drinking water
*7-3 respectively drinking water and wastewater
*8 COD cell tests fully comply with ISO 15705.
Specifications
Specifications
  • Chemical method:
    Chromosulphuric oxidation
  • Pk:
    25 Tests
  • Description:
    COD cell test*<sup>7-1</sup>
  • For unit:
    Prove, Nova 30/60, Multy, Move, Picco
  • Range:
    10 - 150 mg/L
  • Cat. No.:
    1.14540.0001
  • M-Clarity quality level:
    MQ100
Specifications
Accessories
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • Aborption cone for use with COD test for seawater 1.17059.0001
    Aborption cone for use with COD test for seawater 1.17059.0001
    1.15955.0001
    1
    Log in for availability
    1 * 1 ST - 228,00 €
Product Information
  • Catalog #
    Pack type
    Pk
    Availability
    Price
  • 1.06733.0501
    Plastic bottle
    500 g
    Log in for availability
    1 * 500 g