0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
Orders Link
ContactUsLinkComponent
Cap Inserts
CapInsert™ cap inserts for Cryovial® tubes
CapInsert™ cap inserts for Cryovial® tubes
Orange
479-4539
:
Simport Scientific
CapInsert™ cap inserts for Cryovial® tubes
479-4539
:
Simport Scientific
8
CapInsert™ cap inserts for Cryovial® tubes
Orange
37,50 €
1 * 500 ST
-
+
CoC_SIMPT312-10_C19282632
Colour:
Orange
Seq:
05