Order Entry
Austria
Orders LinkContactUsLinkComponent
Scintillation vials

Scintillation vials

20,00 ml, 26×61 mm, Yes, Bulk
Catalog # 720-0496
Supplier:   Simport Scientific
Scintillation vials
Catalog # 720-0496
Supplier:  Simport Scientific
Scintillation vials
Scintillation vials

20,00 ml, 26×61 mm, Yes, Bulk

Price
190,00 €
Log in for availability
1 * 500 ST
Product Details & Documents

These scintillation vials are made of PP. Caps are made of HDPE.

  • Easy removal of caps (¼ turn-twist), leakproof
  • High degree of resistance to organic solvents
  • Shoulderless vial features a full width opening of 23 mm for ease of access
  • Sufficient translucence so that reagent levels or the presence of filter paper can be easily detected

The quality of the sealing system is such that it is not necessary to tighten the closure with a lot of pressure to achieve a leakproof seal. The HDPE closure may not be autoclaved.

Specifications
  • Cap included:
    Yes
  • Ø×H:
    26×61 mm
  • Packed:
    Bulk
  • Capacity:
    20,00 ml
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.