Orders LinkContactUsLinkComponent
Embedding cassettes, M480
Embedding cassettes, M480
White
Catalog # 720-0832
Supplier:   Simport Scientific
Embedding cassettes, M480
Catalog # 720-0832
Supplier:  Simport Scientific
Embedding cassettes, M480
Embedding cassettes, M480
White
Price
213,00 €
Log in for availability
1 * 1.500 ST
Product Details & Documents
Disposable plastic tissue cassettes are suitable for holding and identifying tissue samples in processing, embedding, and sectioning procedures.
  • Resistant to all commonly used histological solvents
  • Designed to accept standard metal lids (M481)
  • Slanted writing surface (45° angle) accepts markings easily
Different colours available.
Specifications
  • Colour:
    White
  • Seq:
    05
Accessories
Product Information
  • Catalog #
    Colour
    Availability
    Price
  • 720-0403
    Silver
    Log in for availability
    1 * 25 ST - 110,00 €