Orders LinkContactUsLinkComponent
pH indicator papers, MQuant®, Supelco®
pH indicator papers, MQuant®, Supelco®
Universal indicator replacement rolls only*, 1 - 10, 1
Catalog # 1.09527.0001
Supplier:   Merck
pH indicator papers, MQuant®, Supelco®
Catalog # 1.09527.0001
Supplier:  Merck
pH indicator papers, MQuant®, Supelco®
pH indicator papers, MQuant®, Supelco®
Universal indicator replacement rolls only*, 1 - 10, 1
Price
66,00 €
Log in for availability
1 * 6 Rolle
Product Details & Documents
Traditional pH indicator test papers, supplied on a 4,8 metre roll, consisting of high quality filter paper impregnated with indicator and mixed indicator solutions then dried and cut to size.
  • Roll format are easy to use and most common ranges are available in a refill format
  • Quality assurance - tests are checked and calibrated against certified buffer solutions
  • Wide and narrow pH ranges available
* no colour scale card supplied
** pack of 3×100 strips
Specifications
  • pH gradation:
    1
  • pH range:
    1 - 10
  • Description:
    Universal indicator replacement rolls only*
  • Cat. No.:
    1.09527.0001
  • M-Clarity quality level:
    MQ100