Orders LinkContactUsLinkComponent
Micro control valves
Micro control valves
Stainless steel manual on/off valve (SMOV), "L" single outlet, connection 1,6 mm, temperature 100 °C, 700 psi pressure
Catalog # SGEA1236283
Micro control valves
Catalog # SGEA1236283
Micro control valves
Stainless steel manual on/off valve (SMOV), "L" single outlet, connection 1,6 mm, temperature 100 °C, 700 psi pressure
Price
204,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Micro control valves provide precision metering and on-off control of gas flows. They are designed specifically for chromatography applications and used at elevated temperatures from high vacuum to medium pressure.
Actuated remotely with a toggle switch or solenoid valve, and a 50 to 55 psi air supply.
Specifications
  • Description:
    Stainless steel manual on/off valve (SMOV), "L" single outlet, connection 1,6 mm, temperature 100 °C, 700 psi pressure
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Seal seat removal tool
    Seal seat removal tool
    SGEA1236101
    Log in for availability
    1 * 1 ST - 20,90 €
Product Information
  • Catalog #
    Availability
    Price
  • VSV-6 Vespel valve seat
    VSV-6 Vespel valve seat
    SGEA123681
    Log in for availability
    1 * 5 ST - 143,00 €
Product Information
  • Catalog #
    Availability
    Price
  • PTFE seals for MOVP valves
    PTFE seals for MOVP valves
    SGEA123687
    Log in for availability
    1 * 2 ST - 32,60 €