Orders LinkContactUsLinkComponent
Micro control valves
Manual on/off valve, pneumatic (MOVPT), "T" dual outlet, stem length 50 mm, connection 1,6 mm, temperature 300 °C, vacuum rated, 700 psi pressure
  SGEA1236103
Micro control valves
  SGEA1236103
Micro control valves
Manual on/off valve, pneumatic (MOVPT), "T" dual outlet, stem length 50 mm, connection 1,6 mm, temperature 300 °C, vacuum rated, 700 psi pressure
813,00 €
1 * 1 ST
Micro control valves provide precision metering and on-off control of gas flows. They are designed specifically for chromatography applications and used at elevated temperatures from high vacuum to medium pressure.
Actuated remotely with a toggle switch or solenoid valve, and a 50 to 55 psi air supply.
  • Description:
    Manual on/off valve, pneumatic (MOVPT), "T" dual outlet, stem length 50 mm, connection 1,6 mm, temperature 300 °C, vacuum rated, 700 psi pressure
Accessories
  •  
    Description
  • SGEA1236101
    Seal seat removal tool
    1 * 1 ST - 20,90 €
  •  
    Description
  • SGEA123681
    VSV-6 Vespel valve seat
    1 * 5 ST - 143,00 €
  •  
    Description
  • SGEA123687
    PTFE seals for MOVP valves
    1 * 2 ST - 32,60 €