Order Entry
Austria
Orders LinkContactUsLinkComponent
Urease

Urease

25 g
Catalog # 1.08489.0025
Supplier:   Merck
Urease
Catalog # 1.08489.0025
Supplier:  Merck
CAS Number:  
CAS Number:  
Price
800,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Urease is used for biochemical research and analysis.

Documents

Specifications

Specifications

  • Pk:
    25 g
  • Cat. No.:
    1.08489.0025
  • Enzyme activity:
    ≥3.0 u/mg
  • Form:
    Light yellow lyophilized powder
  • pH:
    6 - 7
  • Enzyme name:
    Urease

Specifications