Order Entry
Austria
Orders LinkContactUsLinkComponent
Selenium, Supelco®

Selenium, Supelco®

Plastic bottle, 250 g
Catalog # 1.07714.0250
Toxic
Health
Supplier:   Merck
Selenium, Supelco®
Catalog # 1.07714.0250
Supplier:  Merck
CAS Number:  7782-49-2
Selenium, Supelco®

Plastic bottle, 250 g

CAS Number:  7782-49-2
Price
854,00 €
Log in for availability
1 * 250 g
Product Details & Documents
  • CAS Number:7782-49-2
  • Formula:Se
  • MDL Number:MFCD00134090
  • Molecular Weight:78.96 g/mol
  • Merck Index:12,08572
  • Boiling Pt:∼685 °C (1013 hPa)
  • Melting Pt:217 °C
  • Density:4.28 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-957-4
  • ADR:6.1,III
  •  :M-Clarity™ quality level = MQ300
  • UN:3288

Documents

Certificates
Search by Lot Number
Specifications

Specification Test Results

  • Solubility:
    (20 °C) slightly soluble
  • Melting Point:
    217 °C
  • Molar Mass:
    78.96 g/mol
  • Bulk Density:
    1270 kg/m3
  • Boiling Point:
    685 °C (1013 hPa)
  • Vapor Pressure:
    0.001 hPa (20 °C)

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g

Specifications