Orders LinkContactUsLinkComponent
Base materials
Base materials
Peptone from gelatin, pancreatic, 1 kg
Catalog # 1.07284.1000
Supplier:   Merck
Base materials
Catalog # 1.07284.1000
Supplier:  Merck
CAS Number:  
Base materials
Peptone from gelatin, pancreatic, 1 kg
CAS Number:  
Price
360,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Merck products offer high quality for applications in the microbiology field.
  • All products from one manufacturer
  • Constant quality and low costs as a result of bulk buying
  • Custom manufacture following customer developed protocols with excellent quality control
According DIN ISO 9001-2000 and ISO 11133.
Certifications: According DIN ISO 9001-2000 and ISO 11133.
Specifications
Specifications
  • Pk:
    1 kg
  • Description:
    Peptone from gelatin, pancreatic