Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Standard reference materials and buffer solutions, CertiPUR®, Supelco®

Potassium hydrogen phthalate, 4,005 (25°C), Plastic bottle, 5×100 ml, 5
  1.07200.0105
 :   Merck
Standard reference materials and buffer solutions, CertiPUR®, Supelco®
  1.07200.0105
 :  Merck
 :  
Standard reference materials and buffer solutions, CertiPUR®, Supelco®

Potassium hydrogen phthalate, 4,005 (25°C), Plastic bottle, 5×100 ml, 5

 :  
 
171,00 €
   
1 * 5 ST
 

Materials for the precise calibration and monitoring of pH instruments.

  • Traceable to NIST primary standard and PTB
  • Maximum deviation ±0,003 pH units
  • Measured in an independent accredited calibration laboratory for the pH value measurement (DKD-K-14301)
 

 

  • Pack type:
    Plastic bottle
  • Pk:
    5
  • Description:
    Potassium hydrogen phthalate
  • Pk Info:
    5×100 ml
  • pH value:
    4,005 (25°C)
  • Cat. No.:
    1.07200.0105
  • M-Clarity quality level:
    MQ300