Order Entry
Austria
Orders LinkContactUsLinkComponent
Bath fluids, Supelco®

Bath fluids, Supelco®

Oil bath fluid, ≤250 °C, ~5000 cP, 1 l
Catalog # 1.06900.1000
Supplier:   Merck
Bath fluids, Supelco®
Catalog # 1.06900.1000
Supplier:  Merck
CAS Number:  
Bath fluids, Supelco®

Oil bath fluid, ≤250 °C, ~5000 cP, 1 l

CAS Number:  
Price
83,50 €
Log in for availability
1 * 1 L
Product Details & Documents

Bath fluids for improved thermostat life.

  • Oil bath fluid can be used up to approximately 250 °C
  • Heating bath fluid can be used up to approximately 170 °C
  • Non corrosive
  • High flash point
  • Low evaporation

Note: At temperatures approaching the recommended maximum operating temperature it may be advisable to use these products in a fume cupboard or vented area.

Caution: Note: At temperatures approaching the recommended maximum operating temperature it may be advisable to use these products in a fume cupboard or vented area.
1 cP = 1 mPa.s

Specifications

Specifications

  • Pk:
    1 l
  • Description:
    Oil bath fluid
  • Temperature range:
    ≤250 °C
  • Viscosity:
    ~5000 cP
  • Cat. No.:
    1.06900.1000
  • M-Clarity quality level:
    MQ100

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.