Order Entry
Austria
Orders LinkContactUsLinkComponent
Standard reference materials and buffer solutions, CertiPUR®, Supelco®

Standard reference materials and buffer solutions, CertiPUR®, Supelco®

Potassium hydrogen tartrate, 3,639 (25°C), Glass bottle for solids, 1×25 g, 25 g
Catalog # 1.01963.0025
Supplier:   Merck
Standard reference materials and buffer solutions, CertiPUR®, Supelco®
Catalog # 1.01963.0025
Supplier:  Merck
CAS Number:  
Standard reference materials and buffer solutions, CertiPUR®, Supelco®

Potassium hydrogen tartrate, 3,639 (25°C), Glass bottle for solids, 1×25 g, 25 g

CAS Number:  
Price
199,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Materials for the precise calibration and monitoring of pH instruments.

  • Traceable to NIST primary standard and PTB
  • Maximum deviation ±0,003 pH units
  • Measured in an independent accredited calibration laboratory for the pH value measurement (DKD-K-14301)
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Description:
    Potassium hydrogen tartrate
  • Pk Info:
    1×25 g
  • pH value:
    3,639 (25°C)
  • Cat. No.:
    1.01963.0025
  • M-Clarity quality level:
    MQ100

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.