LAMPE HALOGENE PRE-ALIGN., CERTIF. SECOM 1 * 1 items
Catalog # SECO80ZZ0034
Supplier: AQUALABO ANALYSE
LAMPE HALOGENE PRE-ALIGN., CERTIF. SECOM 1 * 1 items
Catalog # SECO80ZZ0034
Supplier: AQUALABO ANALYSE
Frequently Bought Together
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Glove liners, ActivArmr® 78-110
Supplier: Ansell
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
ESD safety shoes, slip-on, X-Light, 7131032/7131037
Supplier: Abeba
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection