Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Jars, multipurpose, with covers, Nalgene®

2200 ml, 133 mm, 195 mm
  216-8262
   Low Manufacturing Impact
Jars, multipurpose, with covers, Nalgene®
  216-8262
   Low Manufacturing Impact
Jars, multipurpose, with covers, Nalgene®
Jars, multipurpose, with covers, Nalgene®

2200 ml, 133 mm, 195 mm

 
108,00 €
   
1 * 1 ST
 

Polycarbonate, clear.

  • Drill, tap or apply clamps to jars to attach heating or cooling equipment
  • Very durable
  • Autoclavable

Multipurpose jars with cover, which can be used as water baths, aquaria/terraria or freeze-drying chambers.

 
  • Ext.Ø:
    133 mm
  • Capacity:
    2200 ml
  • Height:
    195 mm
  • Cat. No.:
    216-8262
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact