Orders LinkContactUsLinkComponent
Dropping bottles, PP, Nalgene®
Dropping bottles, PP, Nalgene®
250 ml
Catalog # NALGDS2420-0250
   Low Manufacturing Impact
Dropping bottles, PP, Nalgene®
Catalog # NALGDS2420-0250
   Low Manufacturing Impact
Dropping bottles, PP, Nalgene®
Dropping bottles, PP, Nalgene®
250 ml
Price
73,60 €
Log in for availability
1 * 6 ST
Product Details & Documents
PPCO with non-drip closure and cap made from blue PP.
  • Autoclavable even when full with tips uncapped
  • Leakproof closure
  • Integral non-drip closure with attached cap
Ideal for sterile solutions.
Specifications
  • Capacity:
    250 ml
  • Cat. No.:
    NALGDS2420-0250
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact